Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
tps10
Entrez Gene ID
732751
Gene Name
terpene synthase10
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112380.1 NP_001105850.1 - 1059 96
GenBank mRNA AY928079.1 - - 1059 96

View Full Product Details