Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
smt2
Entrez Gene ID
100192647
Gene Name
sterol methyl transferase2
Gene Aliases
ZEAMMB73_766247
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001137859.2 NP_001131331.2 - 1451 62
GenBank mRNA BT034335.1 - - 1190 62
BT034685.2 - - 1451 62
BT040017.1 - - 1156 62
BT066549.1 - - 1290 62

View Full Product Details