Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cax1
Entrez Gene ID
541851
Gene Name
calcium exchanger1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111529.1 NP_001104999.1 - 238 76
GenBank mRNA AF256229.1 - - 238 76

View Full Product Details