Assay Design
The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Gene Transcripts
- Gene Symbol
- hmg2
- Entrez Gene ID
-
541951
- Gene Name
- high mobility group box protein2
- Gene Aliases
-
nfd104
- Chromosome Location
-
-
- Gene Symbol
- LOC100382741
- Entrez Gene ID
-
100382741
- Gene Name
- uncharacterized LOC100382741
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BT034240.1 |
- |
- |
378 |
89 |
BT041105.1 |
- |
- |
393 |
89 |
BT054508.1 |
- |
- |
332 |
89 |
View Full Product Details