Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
mlo2
Entrez Gene ID
542061
Gene Name
barley mlo defense gene homolog2
Gene Aliases
-Mlo2, ZEAMMB73_604520, Zm-Mlo2
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111698.1 NP_001105168.1 - 244 63
GenBank mRNA AY029313.1 - - 244 63
AY105309.1 - - 244 63
BT063312.1 - - 275 63

View Full Product Details