Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
mlo8
Entrez Gene ID
542065
Gene Name
barley mlo defense gene homolog8
Gene Aliases
-Mlo8, ZEAMMB73_016515, Zm-Mlo8
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111701.1 NP_001105171.1 - 748 108
GenBank mRNA AY029319.1 - - 748 108
AY104078.1 - - 748 108
BT043122.1 - - 927 108
BT056039.1 - - 820 108
EU971076.1 - - 794 108

View Full Product Details