Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
adh2
Entrez Gene ID
542364
Gene Name
alcohol dehydrogenase2
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111940.1 NP_001105410.1 - 830 85
GenBank mRNA AY104300.2 - - 825 85
BT041397.1 - - 487 85
X01965.1 - - 830 85

View Full Product Details