Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
dps1
Entrez Gene ID
542379
Gene Name
dihydrodipicolinate synthase1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111955.1 NP_001105425.1 - 252 91
GenBank mRNA X52850.1 - - 252 91

View Full Product Details