Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
fdh1
Entrez Gene ID
542459
Gene Name
formaldehyde dehydrogenase homolog1
Gene Aliases
fdh
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112015.1 NP_001105485.1 - 259 72
GenBank mRNA AY111235.1 - - 259 72
Y11029.1 - - 259 72

View Full Product Details