Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
mlo3
Entrez Gene ID
542508
Gene Name
barley mlo defense gene homolog3
Gene Aliases
-Mlo3, Zm-Mlo3
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112057.1 NP_001105527.1 - 196 83
GenBank mRNA AY029314.1 - - 196 83
AY108219.1 - - 180 83

View Full Product Details