Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ccp1
Entrez Gene ID
542701
Gene Name
cysteine protease1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112215.1 NP_001105685.1 - 494 60
GenBank mRNA AY104312.2 - - 555 60
D45402.1 - - 494 60

View Full Product Details