Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
sps1
Entrez Gene ID
542711
Gene Name
sucrose phosphate synthase1
Gene Aliases
SPS
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112224.1 NP_001105694.1 - 420 82
GenBank mRNA M97550.1 - - 420 82

View Full Product Details