Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
sgo1
Entrez Gene ID
606461
Gene Name
shugoshin centromeric cohesion1
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112321.1 NP_001105791.1 - 524 86
GenBank mRNA AY964185.1 - - 524 86

View Full Product Details