Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
atg6
Entrez Gene ID
732797
Gene Name
beclin 1 protein
Gene Aliases
ZEAMMB73_446364
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112413.1 NP_001105883.1 - 834 104
GenBank mRNA AM075828.1 - - 834 104
EU957714.1 - - 917 104

View Full Product Details