Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
POD1
Entrez Gene ID
100286773
Gene Name
ascorbate peroxidase
Gene Aliases
ZEAMMB73_012020
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159274.1 NP_001152746.1 - 1050 74
GenBank mRNA BT039906.1 - - 883 74
EU973991.1 - - 1050 74

View Full Product Details