Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cesa2
Entrez Gene ID
542564
Gene Name
cellulose synthase2
Gene Aliases
CesA-2, ZEAMMB73_144604
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112104.1 NP_001105574.1 - 3552 98
GenBank mRNA AF200526.1 - - 3552 98

View Full Product Details