Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
sod3
Entrez Gene ID
542764
Gene Name
superoxide dismutase3
Gene Aliases
SODA.4
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112272.1 NP_001105742.1 - 953 101
GenBank mRNA M33119.1 - - 953 101
X12540.1 - - 953 101

View Full Product Details