Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Atg8d
Entrez Gene ID
100240697
Gene Name
autophagy-related 8d
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144023.1 NP_001137495.1 - 850 98
GenBank mRNA FJ445010.1 - - 850 98

View Full Product Details