Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
pip2b
Entrez Gene ID
542644
Gene Name
plasma membrane intrinsic protein2
Gene Aliases
ZEAMMB73_762428
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112168.1 NP_001105638.1 - 351 105
GenBank mRNA AF326492.1 - - 351 105
BT065379.1 - - 326 105
EU967848.1 - - 364 105

View Full Product Details