Gene Transcripts
- Gene Symbol
- Snora17
- Entrez Gene ID
- 100306954
- Gene Name
- small nucleolar RNA, H/ACA box 17
- Gene Aliases
- ACA17
- Chromosome Location
- Chr. 2: 26639190 - 26639321 on Build GRCm38
- UniGene ID
- Mm.491822
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028571.1 |
|
1 |
78 |
|
- Gene Symbol
- Snhg7
- Entrez Gene ID
- 72091
- Gene Name
- small nucleolar RNA host gene 7
- Gene Aliases
- 2610002F03Rik, AI427062
- Chromosome Location
- Chr. 2: 26637176 - 26640244 on Build GRCm38
- UniGene ID
- Mm.27103
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC046478.1 |
- |
3 |
393 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CJ213652.1 |
- |
2 |
335 |
|
Chr. 2 - 26639188 - 26640164 [-] |
View Full Product Details