Gene Transcripts
- Gene Symbol
- Fate1
- Entrez Gene ID
- 77905
- Gene Name
- fetal and adult testis expressed 1
- Gene Aliases
- 9230118O15Rik
- Chromosome Location
- Chr. X: 71972986 - 71989046 on Build GRCm38
- UniGene ID
- Mm.99811
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003243.2 |
|
2 |
111 |
|
GenBank mRNA |
AK020359.1 |
- |
2 |
112 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY721172.1 |
- |
2 |
110 |
|
Chr. X - 71972987 - 71988731 [+] |
View Full Product Details