Gene Transcripts
- Gene Symbol
- Mgarp
- Entrez Gene ID
- 67749
- Gene Name
- mitochondria localized glutamic acid rich protein
- Gene Aliases
- 4930583H14Rik, AI195347, CESP-1, HUMMR, Osap, Qsap
- Chromosome Location
- Chr. 3: 51388412 - 51396547 on Build GRCm38
- UniGene ID
- Mm.273339
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028121.1 |
|
1 |
139 |
|
GenBank mRNA |
BC022740.1 |
- |
1 |
94 |
|
View Full Product Details