Gene Transcripts
- Gene Symbol
- Mir99ahg
- Entrez Gene ID
- 77994
- Gene Name
- Mir99a and Mirlet7c-1 host gene (non-protein coding)
- Gene Aliases
- 2810055G20Rik, D730043B02Rik
- Chromosome Location
- Chr. 16: 77329328 - 77558428 on Build GRCm38
- UniGene ID
- Mm.394688
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB656236.1 |
- |
2 |
375 |
|
Chr. 16 - 77329332 - 77501605 [+] |
GenBankmRNA |
CF553191.1 |
- |
2 |
498 |
|
Chr. 16 - 77329211 - 77594731 [+] |
GenBankmRNA |
BB655639.1 |
- |
4 |
432 |
|
Chr. 16 - 77236319 - 77360897 [+] |
GenBankmRNA |
BB647956.1 |
- |
4 |
434 |
|
Chr. 16 - 77236317 - 77501580 [+] |
GenBankmRNA |
BB663466.1 |
- |
4 |
432 |
|
Chr. 16 - 77236319 - 77596954 [+] |
View Full Product Details