Gene Transcripts
- Gene Symbol
- Vmn1r-ps79
- Entrez Gene ID
- 100042966
- Gene Name
- vomeronasal 1 receptor, pseudogene 79
- Gene Aliases
- Gm652
- Chromosome Location
- Chr. 7: 20356626 - 20358918 on Build GRCm38
- UniGene ID
- Mm.431911
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_030707.1 |
|
1 |
251 |
|
GenBank mRNA |
BC115870.1 |
- |
1 |
152 |
|
View Full Product Details