Gene Transcripts

Gene Symbol
Smok4a
Entrez Gene ID
272667
Gene Name
sperm motility kinase 4A
Gene Aliases
4930513D10Rik, Smok2c
Chromosome Location
Chr. 17: 13521456 - 13528434 on Build GRCm38
UniGene ID
Mm.444718
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_030763.1 2 1183
GenBank mRNA AK029728.1 - 2 1184
BC125398.1 - 1 742
BC132092.1 - 1 742

View Full Product Details