Gene Transcripts
- Gene Symbol
- Speer8-ps1
- Entrez Gene ID
- 74062
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 8, pseudogene 1
- Gene Aliases
- 4933405L09Rik, SPEER-8, Speer8-ps
- Chromosome Location
- Chr. 5: 14945294 - 14978541 on Build GRCm38
- UniGene ID
- Mm.394766
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001584.3 |
|
2 |
62 |
|
- Gene Symbol
- Gm38698
- Entrez Gene ID
- 105242428
- Gene Name
- predicted gene, 38698
- Gene Aliases
- Chromosome Location
- Chr. 5: 15511563 - 15550662 on Build GRCm38
- UniGene ID
- Mm.394766
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK133285.1 |
- |
|
127 |
|
View Full Product Details