Gene Transcripts
- Gene Symbol
- Sec14l1
- Entrez Gene ID
- 74136
- Gene Name
- SEC14-like 1 (S. cerevisiae)
- Gene Aliases
- 1200017E04Rik, 2810012L19Rik, Naa-35
- Chromosome Location
- Chr. 11: 117115172 - 117159268 on Build GRCm38
- UniGene ID
- Mm.272312
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029459.1 |
|
3 |
1109 |
|
GenBank mRNA |
BC055849.1 |
- |
3 |
1029 |
|
View Full Product Details