Gene Transcripts

Gene Symbol
Snhg7
Entrez Gene ID
72091
Gene Name
small nucleolar RNA host gene 7
Gene Aliases
2610002F03Rik, AI427062
Chromosome Location
Chr. 2: 26637176 - 26640244 on Build GRCm38
UniGene ID
Mm.27103
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024068.2 4 537
GenBank mRNA AK011283.1 - 4 538
AK028166.1 - 4 538
BC046478.1 - 3 2256
CB245554.1 - 1 245


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY710585.1 - 4 534 Chr. 2 - 26637229 - 26640241 [-]
GenBankmRNA BY710095.1 - 4 532 Chr. 2 - 26637299 - 26640239 [-]
GenBankmRNA BY274561.1 - 4 378 Chr. 2 - 26637331 - 26640241 [-]
GenBankmRNA AA041850.1 - 3 364 Chr. 2 - 26637257 - 26640113 [-]
GenBankmRNA BY281080.1 - 4 378 Chr. 2 - 26637366 - 26640241 [-]

View Full Product Details