Gene Transcripts
- Gene Symbol
- Snhg7
- Entrez Gene ID
- 72091
- Gene Name
- small nucleolar RNA host gene 7
- Gene Aliases
- 2610002F03Rik, AI427062
- Chromosome Location
- Chr. 2: 26637176 - 26640244 on Build GRCm38
- UniGene ID
- Mm.27103
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY710585.1 |
- |
4 |
405 |
|
Chr. 2 - 26637229 - 26640241 [-] |
GenBankmRNA |
BY710095.1 |
- |
4 |
403 |
|
Chr. 2 - 26637299 - 26640239 [-] |
GenBankmRNA |
CA466784.1 |
- |
4 |
445 |
|
Chr. 2 - 26637217 - 26640244 [-] |
GenBankmRNA |
AA041850.1 |
- |
3 |
235 |
|
Chr. 2 - 26637257 - 26640113 [-] |
GenBankmRNA |
AV572272.1 |
- |
4 |
393 |
|
Chr. 2 - 26637440 - 26640229 [-] |
View Full Product Details