Gene Transcripts
- Gene Symbol
- Scarna3b
- Entrez Gene ID
- 100217447
- Gene Name
- small Cajal body-specific RNA 3B
- Gene Aliases
- MBI-100
- Chromosome Location
- Chr. 12: 80391614 - 80391737 on Build GRCm38
- UniGene ID
- Mm.453532
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028544.1 |
|
1 |
103 |
|
GenBank mRNA |
AF357403.1 |
- |
1 |
103 |
|
- Gene Symbol
- Mir1843a
- Entrez Gene ID
- 100526530
- Gene Name
- microRNA 1843a
- Gene Aliases
- Mir1843, mir-1843a, mmu-mir-1843, mmu-mir-1843a
- Chromosome Location
- Chr. 12: 80391613 - 80391677 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037207.1 |
|
1 |
43 |
|
View Full Product Details