Gene Transcripts

Gene Symbol
Prr18
Entrez Gene ID
320111
Gene Name
proline rich 18
Gene Aliases
9630019K15Rik
Chromosome Location
Chr. 17: 8340406 - 8344113 on Build GRCm38
UniGene ID
Mm.442510
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028280.1 1 46
GenBank mRNA AK045379.1 - 1 22
BC060190.1 - 1 46 6833275

View Full Product Details