Gene Transcripts

Gene Symbol
Thap6
Entrez Gene ID
381650
Gene Name
THAP domain containing 6
Gene Aliases
4930534K21Rik, AI448650
Chromosome Location
Chr. 5: 91962882 - 91972137 on Build GRCm38
UniGene ID
Mm.159453
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028429.1 5 1552
GenBank mRNA AK039179.1 - 4 1428
AK080650.1 - 4 1427
BC089377.1 - 5 1101

View Full Product Details