Gene Transcripts
- Gene Symbol
- Speer5-ps1
- Entrez Gene ID
- 70365
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 5, pseudogene 1
- Gene Aliases
- 1700007C10Rik, SPEER-5, Speer5-ps
- Chromosome Location
- Chr. 10: 44170447 - 44219585 on Build GRCm38
- UniGene ID
- Mm.87073
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001582.2 |
|
7 |
1223 |
|
NR_027506.1 |
|
4 |
761 |
|
GenBank mRNA |
BB679670.1 |
- |
1 |
204 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV206592.1 |
- |
1 |
100 |
|
Chr. 10 - 44219277 - 44219583 [+] |
GenBankmRNA |
AV039433.2 |
- |
1 |
110 |
|
Chr. 10 - 44219303 - 44219583 [+] |
GenBankmRNA |
AV210376.1 |
- |
1 |
135 |
|
Chr. 10 - 44219278 - 44219583 [+] |
View Full Product Details