Gene Transcripts
- Gene Symbol
- Btbd19
- Entrez Gene ID
- 78611
- Gene Name
- BTB (POZ) domain containing 19
- Gene Aliases
- 9530048O09Rik, BB115071
- Chromosome Location
- Chr. 4: 117119218 - 117125725 on Build GRCm38
- UniGene ID
- Mm.446227
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024078.1 |
|
7 |
1388 |
|
GenBank mRNA |
AK141215.1 |
- |
2 |
620 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
NM_001033187.1 |
- |
|
620 |
|
Chr. - - [-] |
View Full Product Details