Gene Transcripts
- Gene Symbol
- Rnf44
- Entrez Gene ID
- 105239
- Gene Name
- ring finger protein 44
- Gene Aliases
- AI854545, mKIAA1100
- Chromosome Location
- Chr. 13: 54679399 - 54693995 on Build GRCm38
- UniGene ID
- Mm.25366
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027395.1 |
|
1 |
103 |
|
NR_027396.1 |
|
1 |
103 |
|
GenBank mRNA |
BY346776.1 |
- |
1 |
72 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CJ176406.1 |
- |
1 |
26 |
|
Chr. 13 - 54683717 - 54693882 [-] |
View Full Product Details