Gene Transcripts
- Gene Symbol
- Cyp2d13
- Entrez Gene ID
- 68444
- Gene Name
- cytochrome P450, family 2, subfamily d, polypeptide 13
- Gene Aliases
- 1300007K12Rik
- Chromosome Location
- Chr. 15: 82636750 - 82642045 on Build GRCm38
- UniGene ID
- Mm.443212
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003552.1 |
|
4 |
697 |
|
GenBank mRNA |
AK004933.1 |
- |
|
527 |
|
BC018344.1 |
- |
4 |
677 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI147110.1 |
- |
1 |
120 |
|
Chr. 15 - 82636826 - 82640267 [-] |
GenBankmRNA |
BI144949.1 |
- |
4 |
653 |
|
Chr. 15 - 82639353 - 82642001 [-] |
View Full Product Details