Gene Transcripts
- Gene Symbol
- Snora35
- Entrez Gene ID
- 100303736
- Gene Name
- small nucleolar RNA, H/ACA box 35
- Gene Aliases
- MBI-36
- Chromosome Location
- Chr. X: 146994390 - 146994508 on Build GRCm38
- UniGene ID
- Mm.453529
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028446.1 |
|
1 |
23 |
|
GenBank mRNA |
AF357423.1 |
- |
1 |
23 |
|
View Full Product Details