Gene Transcripts
- Gene Symbol
- Mirlet7d
- Entrez Gene ID
- 387247
- Gene Name
- microRNA let7d
- Gene Aliases
- Mirnlet7d, let-7d, mmu-let-7d
- Chromosome Location
- Chr. 13: 48536012 - 48536114 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029656.1 |
|
1 |
35 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK160312.1 |
- |
|
1040 |
|
Chr. - - [-] |
View Full Product Details