Gene Transcripts
- Gene Symbol
- Fbxo34
- Entrez Gene ID
- 78938
- Gene Name
- F-box protein 34
- Gene Aliases
- 2900057B08Rik, 5830426G16Rik
- Chromosome Location
- Chr. 14: 47472561 - 47531962 on Build GRCm38
- UniGene ID
- Mm.11935
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027414.1 |
|
2 |
251 |
|
GenBank mRNA |
AK013711.1 |
- |
2 |
188 |
|
AK171630.1 |
- |
3 |
255 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY712688.1 |
- |
2 |
184 |
|
Chr. 14 - 47472628 - 47529410 [+] |
GenBankmRNA |
CA542265.1 |
- |
3 |
237 |
|
Chr. 14 - 47472651 - 47526236 [+] |
View Full Product Details