Gene Transcripts
- Gene Symbol
- Rnu11
- Entrez Gene ID
- 353373
- Gene Name
- U11 small nuclear RNA
- Gene Aliases
- MBII-407, Mirn801, mir-801, mmu-mir-801
- Chromosome Location
- Chr. 4: 132270079 - 132270186 on Build GRCm38
- UniGene ID
- Mm.424675
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002865.2 |
|
1 |
39 |
|
GenBank mRNA |
AF357361.1 |
- |
1 |
39 |
|
View Full Product Details