Gene Transcripts
- Gene Symbol
- Dlx1as
- Entrez Gene ID
- 111970
- Gene Name
- distal-less homeobox 1, antisense
- Gene Aliases
- Dix1as, Dlx1os
- Chromosome Location
- Chr. 2: 71530638 - 71537891 on Build GRCm38
- UniGene ID
- Mm.4543
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002854.2 |
|
2 |
1922 |
|
GenBank mRNA |
AK132348.1 |
- |
2 |
1923 |
|
View Full Product Details