Gene Transcripts
- Gene Symbol
- Tspy-ps
- Entrez Gene ID
- 22109
- Gene Name
- testis specific protein-Y encoded, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. Y: 1055764 - 1058868 on Build GRCm38
- UniGene ID
- Mm.10106
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027507.1 |
|
2 |
80 |
|
GenBank mRNA |
AJ001378.1 |
- |
2 |
82 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB013038.2 |
- |
1 |
409 |
|
Chr. Y - 1055764 - 1057642 [-] |
View Full Product Details