Gene Transcripts
- Gene Symbol
- Vaultrc5
- Entrez Gene ID
- 378472
- Gene Name
- vault RNA component 5
- Gene Aliases
- Gm22653, Mvg1
- Chromosome Location
- Chr. 18: 36801763 - 36802107 on Build GRCm38
- UniGene ID
- Mm.482379
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027885.1 |
|
1 |
36 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF210457.1 |
- |
1 |
36 |
|
Chr. - - [-] |
View Full Product Details