Gene Transcripts

Gene Symbol
4931409K22Rik
Entrez Gene ID
231045
Gene Name
RIKEN cDNA 4931409K22 gene
Gene Aliases
Iqca1l, Iqca1p1
Chromosome Location
Chr. 5: 24543434 - 24555469 on Build GRCm38
UniGene ID
Mm.255800
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033188.1 6 1222
GenBank mRNA AK029839.1 - 6 1164
BC137999.1 - 3 453 8861018
BC145792.1 - 3 453 40131095

View Full Product Details