Gene Transcripts
- Gene Symbol
- Npm3-ps1
- Entrez Gene ID
- 108176
- Gene Name
- nucleoplasmin 3, pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 85076141 - 85077126 on Build GRCm38
- UniGene ID
- Mm.431982
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002702.1 |
|
1 |
18 |
|
View Full Product Details