Gene Transcripts
- Gene Symbol
- LINC00526
- Entrez Gene ID
- 147525
- Gene Name
- long intergenic non-protein coding RNA 526
- Gene Aliases
- C18orf18, HsT959
- Chromosome Location
- Chr. 18: 5236724 - 5238029 on Build GRCh38
- UniGene ID
- Hs.657197
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026849.1 |
|
1 |
888 |
|
GenBank mRNA |
AK292743.1 |
- |
1 |
1465 |
|
BC010538.1 |
- |
1 |
888 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM714979.1 |
- |
1 |
200 |
|
Chr. 18 - 5236718 - 5237341 [-] |
GenBankmRNA |
BQ067788.1 |
- |
|
54 |
|
Chr. - - [-] |
GenBankmRNA |
BX431478.2 |
- |
1 |
32 |
|
Chr. 18 - 5236724 - 5237173 [-] |
GenBankmRNA |
BM716520.1 |
- |
1 |
200 |
|
Chr. 18 - 5236718 - 5237341 [-] |
GenBankmRNA |
BX431479.2 |
- |
1 |
35 |
|
Chr. 18 - 5236724 - 5237172 [-] |
View Full Product Details