Gene Transcripts

Gene Symbol
PSORS1C3
Entrez Gene ID
100130889
Gene Name
psoriasis susceptibility 1 candidate 3 (non-protein coding)
Gene Aliases
NCRNA00196
Chromosome Location
Chr. 6: 31173735 - 31177899 on Build GRCh38
UniGene ID
Hs.670091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026816.1 3 311
GenBank mRNA AB932952.1 - 2 278
AB932954.1 - 3 309
AB933312.1 - 4 576
AB933313.1 - 4 342
AB933320.1 - 6 794
AB933321.1 - 4 490
AY484516.1 - 3 311
LC027944.1 - 3 545
LC027945.1 - 4 603
LC027946.1 - 3 570
LC050981.1 - 5 787
LC050982.1 - 5 868
LC050983.1 - 5 634
LC050984.1 - 3 342
LC050985.1 - 4 463
LC050986.1 - 6 820
LC050987.1 - 7 1087
LC050988.1 - 2 309
LC050989.1 - 4 601
LC050990.1 - 6 673
LC050991.1 - 4 837

View Full Product Details