Gene Transcripts

Gene Symbol
PSORS1C3
Entrez Gene ID
100130889
Gene Name
psoriasis susceptibility 1 candidate 3 (non-protein coding)
Gene Aliases
NCRNA00196
Chromosome Location
Chr. 6: 31173735 - 31177899 on Build GRCh38
UniGene ID
Hs.670091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026816.1 3 550
GenBank mRNA AB932952.1 - 2 517
AB932953.1 - 3 744
AB932954.1 - 3 548
AB933312.1 - 4 815
AB933313.1 - 4 581
AB933320.1 - 6 1033
AB933321.1 - 4 729
AY484516.1 - 3 550
LC027944.1 - 3 784
LC027945.1 - 4 842
LC027946.1 - 3 809
LC050979.1 - 4 640
LC050980.1 - 3 620
LC050981.1 - 5 1026
LC050982.1 - 5 1107
LC050983.1 - 5 873
LC050984.1 - 3 581
LC050985.1 - 4 702
LC050986.1 - 6 1059
LC050987.1 - 7 1326
LC050988.1 - 2 548
LC050989.1 - 4 840
LC050990.1 - 6 912
LC050991.1 - 4 1076

View Full Product Details