Gene Transcripts

Gene Symbol
KLKP1
Entrez Gene ID
606293
Gene Name
kallikrein pseudogene 1
Gene Aliases
KARMA, KLK31P, KRSP1, PsiKLK1, YKLK1
Chromosome Location
Chr. 19: 50882096 - 50896398 on Build GRCh38
UniGene ID
Hs.643424
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002948.1 4 460
GenBank mRNA AF355199.1 - 1 22
AF464754.1 - 1 198
AY533562.1 - 4 460
DQ086829.1 - 1 867
DQ211698.1 - 4 2183

View Full Product Details