Gene Transcripts

Gene Symbol
AFG3L1P
Entrez Gene ID
172
Gene Name
AFG3 like matrix AAA peptidase subunit 1, pseudogene
Gene Aliases
AFG3, AFG3L1
Chromosome Location
Chr. 16: 89972580 - 90000787 on Build GRCh38
UniGene ID
Hs.534773
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003226.1 2 344
NR_003227.1 2 271
NR_003228.1 2 310
GenBank mRNA AF329691.1 - 1 95
AF329693.1 - 1 95
AF329694.1 - 1 95
AK056488.1 - 2 297
AK091293.1 - 2 307
AK097044.1 - 2 258
AK126911.1 - 2 304
DA707802.1 - 2 310


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB157156.1 - 2 304 Chr. 16 - 89972586 - 89981802 [+]
GenBankmRNA DA127983.1 - 2 304 Chr. 16 - 89972586 - 89981861 [+]
GenBankmRNA DA207089.1 - 2 304 Chr. 16 - 89972586 - 89981830 [+]
GenBankmRNA BM553984.1 - 2 276 Chr. 16 - 89972594 - 89978394 [+]
GenBankmRNA DR003672.1 - 2 270 Chr. 16 - 89972627 - 89981857 [+]
GenBankmRNA DB162853.1 - 2 304 Chr. 16 - 89972586 - 89979870 [+]
GenBankmRNA DA009807.1 - 2 258 Chr. 16 - 89972593 - 89981920 [+]
GenBankmRNA DA742209.1 - 2 297 Chr. 16 - 89972593 - 89980282 [+]
GenBankmRNA DA927941.1 - 2 258 Chr. 16 - 89972594 - 89981891 [+]
GenBankmRNA DA740893.1 - 2 256 Chr. 16 - 89972595 - 89981803 [+]
GenBankmRNA DA104810.1 - 2 305 Chr. 16 - 89972587 - 89981843 [+]
GenBankmRNA DR005480.1 - 2 272 Chr. 16 - 89972625 - 89981811 [+]
GenBankmRNA DA474196.1 - 2 307 Chr. 16 - 89972617 - 89981824 [+]
GenBankmRNA DA112780.1 - 2 302 Chr. 16 - 89972589 - 89981926 [+]
GenBankmRNA DA114236.1 - 2 323 Chr. 16 - 89972587 - 89981818 [+]
GenBankmRNA BG771795.1 - 2 247 Chr. 16 - 89972648 - 89979609 [+]
GenBankmRNA DA586987.1 - 2 304 Chr. 16 - 89972586 - 89980309 [+]
GenBankmRNA HY066220.1 - 2 293 Chr. 16 - 89972597 - 89980302 [+]
GenBankmRNA DA198586.1 - 2 304 Chr. 16 - 89972586 - 89980332 [+]
GenBankmRNA DB153965.1 - 2 304 Chr. 16 - 89972586 - 89980337 [+]
GenBankmRNA DA857528.1 - 2 298 Chr. 16 - 89972593 - 89981813 [+]
GenBankmRNA DA419322.1 - 2 297 Chr. 16 - 89972594 - 89980335 [+]
GenBankmRNA DB164996.1 - 2 295 Chr. 16 - 89972595 - 89981861 [+]
GenBankmRNA DB181830.1 - 2 270 Chr. 16 - 89972581 - 89980337 [+]
GenBankmRNA DN992972.1 - 2 199 Chr. 16 - 89972621 - 89981865 [+]
GenBankmRNA BG570268.1 - 2 218 Chr. 16 - 89972597 - 89984754 [+]
GenBankmRNA DB151189.1 - 2 297 Chr. 16 - 89972593 - 89980335 [+]
GenBankmRNA DB069789.1 - 2 305 Chr. 16 - 89972586 - 89981877 [+]
GenBankmRNA HY069034.1 - 2 293 Chr. 16 - 89972597 - 89980329 [+]
GenBankmRNA DA989185.1 - 2 265 Chr. 16 - 89972587 - 89981878 [+]
GenBankmRNA DA511246.1 - 2 305 Chr. 16 - 89972585 - 89981816 [+]
GenBankmRNA DA918429.1 - 2 297 Chr. 16 - 89972593 - 89981833 [+]
GenBankmRNA DA898575.1 - 2 304 Chr. 16 - 89972587 - 89981827 [+]
GenBankmRNA DA387750.1 - 2 308 Chr. 16 - 89972582 - 89981918 [+]
GenBankmRNA DA059398.1 - 2 295 Chr. 16 - 89972595 - 89979448 [+]

View Full Product Details